BBL™ Selective 7H11 (Mitchison) Agar Slants, BD
Catalog # CA-90004-926
Supplier: BD
CAS Number:
Specifications
- Description:BBL™ Selective 7H11 (Mitchison) Agar Slants
- Size:20 x 148 mm
- Cat. no.:CA-90004-926
- Supplier No.:297639
Specifications
About this item
Selective 7H11 Agar Slants are used in qualitative procedures for isolation and cultivation of mycobacteria, especially Mycobacterium tuberculosis, from clinical and nonclinical specimens.
- Antimicrobial agents added to reduce the need for decontamination procedures
- Supplemented pancreatic digest of casein
- Pyruvate enhances the recovery of M. bovis
The Selective 7H11 Agar is 7H11 Agar modified by the addition of four antimicrobial agents: polymyxin B, carbenicillin, amphotericin B, and trimethoprim lactate.