Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Triangular Support Stand

Triangular Support Stand with 8 Legs, 24"
  470202-852
 :  H-21320
Triangular Support Stand
  470202-852
Triangular Support Stand
Triangular Support Stand

Triangular Support Stand with 8 Legs, 24"

 
$86.15
   
Each
 

Bases are cast iron with black enamel finish. Rods are zinc plated steel and threaded to fit into the base.

  • Base legs: 5"
  • Rod: 1/2" x 24"

 : Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.

 
  • Description:
    Triangular Support Stand with 8 Legs
  • Length:
    24"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.