Orders LinkContactUsLinkComponent
Hooked Weight Set
Hooked Weight Set
Economical Stainless Steel Class 7 Hook Weight Set with No Certificate, 10 N - 0.1 N
Catalog # 10754-350
Supplier:   Troemner
Supplier #:  30390785
Hooked Weight Set
Catalog # 10754-350
Supplier:  Troemner
Hooked Weight Set
Hooked Weight Set
Economical Stainless Steel Class 7 Hook Weight Set with No Certificate, 10 N - 0.1 N
Price
$795.16
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
  • Set of nine
  • Variety of sizes
  • Durable plastic case
Units in are in Newtons.
Delivery information: This set includes one 10N unit, one 5N unit, two 2N units, one 1N unit, one 0.5N, two 0.2N units, and one 0.1N unit for a set of nine. It also includes a durable plastic case.
Specifications
  • Description:
    Economical Stainless Steel Class 7 Hook Weight Set with No Certificate
  • Range:
    10 N - 0.1 N