ContactUsLinkComponent
LBP, Soluble ELISA Kit, Enzo Life Sciences
LBP, Soluble ELISA Kit, Enzo Life Sciences
  75786-320
 :  
LBP, Soluble ELISA Kit, Enzo Life Sciences
  75786-320
 :  850-304-KI01
 :  
  • Description:
    LBP, Soluble ELISA Kit
  • Size:
    1 x 96 wells
  • Sensitivity:
    Range 5-50ng/ml
  • Cat. no.:
    75786-320
  • Supplier No.:
    850-304-KI01
Quantitative determination of human, dog, pig, rabbit, bovine and horse-soluble LBP levels.
For the quantitative determination of human, dog, pig, rabbit, bovine and horse soluble LBP levels in serum, plasma and cell culture medium.
 : Contains 1 precoated ELISA module1 vial anti-LBP (human) detecting MAb1 vial LBP standard (human)1 vial reference serum2 tablets PBS1 vial dilution buffer1 vial Tween 201 vial stopping solution1 vial substrate solution.