Order Entry
Canada
ContactUsLinkComponent
Two-Circle Microscope Slides for Fluorescent Microscopy, Springside Scientific
Two-Circle Microscope Slides for Fluorescent Microscopy, Springside Scientific
Catalog # 75836-216
undefined
Two-Circle Microscope Slides for Fluorescent Microscopy, Springside Scientific
Catalog # 75836-216
Supplier Number:  W1030C-C20

Specifications

  • Color:
    White End
  • Description:
    Two-circle slides, charged, 90° corners
  • L×W:
    75×25 mm
  • Thickness:
    1.0 mm
  • Cat. no.:
    75836-216
  • Packaging:
    CS
  • Supplier No.:
    W1030C-C20

Specifications

About this item

Two-Circle Microscope Slides are manufactured from pure white glass, and feature two frosted circles designed for use in fluorescent antibody examination

Slides are standard size. The frosted circles measure 15 mm in diameter and are placed 11 mm apart. Two-circle slides are pre-cleaned, with 90° corners.

Ordering information: Slides are packed 72 per cardboard box. Each box is individually sealed. Each case contains 20 boxes.