Order Entry
Canada
ContactUsLinkComponent
 
Grob Column Test Mix, Restek
Catalog # RK35000
Supplier:  Restek
CAS Number:  
undefined
Grob Column Test Mix, Restek
Catalog # RK35000
Supplier:  Restek
Supplier Number:  35000
CAS Number:  

Specifications

  • Description:
    Grob Test Mix
  • Cat. no.:
    RK35000
  • Supplier No.:
    35000

Specifications

About this item

Grob test mix includes nC10-fatty acid methyl esters (FAME), nC11-FAME, nC12-FAME, 2,3-butanediol, dicyclohexylamine, 2,6-dimethylaniline, 2,6-dimethylphenol, 2-ethylhexanoic acid , nonanal, 1-octanol, undecane (C11) and decane (C10).

  • For use with temperature-programmed conditions
  • Components are in methylene chloride (1 mL/ampoule)