Petri Dish
75838-866
Product available on GSA Advantage®
- Sterility:Sterile
- Description:Petri dish, mono plate
- Dimensions:15×100 mm
- Cat. no.:75838-866
- Packaging:25 per pack
- Supplier No.:330120
Distortion-free petri dishes are optically clear.
- Heavy Duty - reduces breakage during shipping
- Stacking rings - guards against slippage when moving stacked plates
Hardy offers a full selection of petri plates: Mono plate Space Saver Plate Bi-Plate Tri-Plate Quad Plate Membrane Filter Methods Contacts Plate Optically clear polystyrene - for distortion free viewing. Heavy Duty - reduces breakage during shipping. Stacking rings - guards against slippage when moving stacked plates.
Applications: Breweries, wineries and distilleries, cannabis QA testing, cosmetics, environmental, food safety, pharmaceutical manufacturing