Order Entry
Canada
ContactUsLinkComponent
Tryptic Soy Broth (TSB), USP
Tryptic Soy Broth (TSB), USP
Catalog # CA75873-674
CAS Number:  
undefined
Tryptic Soy Broth (TSB), USP
Catalog # CA75873-674
Supplier Number:  U363
CAS Number:  

Specifications

  • Pack type:
    PC bottle
  • Description:
    Tryptic soy broth (TSB), USP
  • Size:
    10/pack
  • Minimum Order Quantity:
    5 packs
  • Cat. no.:
    CA75873-674
  • Application:
    Pharmaceutical Manufacturing
  • Volume:
    490 ml
  • Supplier No.:
    U363

Specifications

About this item

For sterility testing and the cultivation of microorganisms.

Tryptic Soy Broth is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi. Also known as Soybean-Casein Digest Broth, USP.