Standard Methods Agar (Plate Count Agar) with Chloramphenicol, Hardy Diagnostics
10158-954
Product available on GSA Advantage®
:
- Pack type:Plate
- Description:Standard Methods Agar (Plate Count Agar) with Chloramphenicol
- Size:15 x 100 mm
- Minimum Order Quantity:10
- Cat. no.:10158-954
- Supplier No.:W43
For the isolation and enumeration of yeast and mold.
Standard Methods Agar with Chloramphenicol 100ug/L is recommended for the isolation and enumeration of yeast and mold.
: Minimum order of 10 packs