Standard Methods Agar , Hardy Diagnostics
75803-008
Product available on GSA Advantage®
:
- Pack type:Plate
- Description:Standard Methods Agar
- Size:15 x 100mm
- Minimum Order Quantity:10
- Cat. no.:75803-008
- Volume:18 ml
- Supplier No.:G13
For the cultivation and enumeration of microorganisms in dairy products, food, and water.
(Plate Count Agar) For the cultivation and enumeration of microorganisms in dairy products, food, and water.