ContactUsLinkComponent
Hygiena SpotCheck Plus™, Hygiene Non-Instrument Color Tests, Hardy Diagnostics
Hygiena SpotCheck Plus™, Hygiene Non-Instrument Color Tests, Hardy Diagnostics
Catalog # CA10154-026
undefined
Hygiena SpotCheck Plus™, Hygiene Non-Instrument Color Tests, Hardy Diagnostics
Catalog # CA10154-026
Supplier Number:  GL100
Specifications
  • Description:
    SpotCheck Plus™, Hygiene Non-Instrument Color Tests, 100 per box
  • Cat. no.:
    CA10154-026
  • Supplier No.:
    GL100
Specifications
About this item
A rapid non-instrument hygiene monitoring test utilizing the snap-valve™ technology.
  • Detects residual lactose and glucose on surfaces
  • Results in 60 minutes
A rapid non-instrument hygiene monitoring test utilizing the snap-valve™ technology. The reagent in the swab detects residual lactose and glucose on surfaces resulting in a color change. Results in 60 seconds.