CulGenex™ Phosphate Buffered Saline (PBS), 10X
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CulGenex™ Phosphate Buffered Saline (PBS) is recommended for use in various procedures used in biological and molecular research.
Application: Pharmaceutical manufacturing
Specifications
- Storage temperature:2...30 °C
- pH:7.4 - 7.6 ±0.1 at 25 °C
Specifications
Related Information
Certificates
Search by Lot NumberHardy Diagnostics CulGenex
Product Literature
Molecular Product Use Guide
Additional Product Details
Frequently Bought Together