Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Pasteurization Efficacy Kit

Alkaline phosphatase kit, 125 tests/kit, 3×7 ml
Catalog # CA75802-932
 :  Z132
Pasteurization Efficacy Kit
Catalog # CA75802-932
CAS Number:  
Pasteurization Efficacy Kit
Pasteurization Efficacy Kit

Alkaline phosphatase kit, 125 tests/kit, 3×7 ml

CAS Number:  
Price
$536.00
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Alkaline phosphatase is an enzyme naturally produced by mammary cells and present in cow's milk.

  • Naturally produced
  • Concentration may vary
  • Based on Rutgers method
  • Industry: Clinical

The concentration of alkaline phosphatase can vary based on feed, season, breed of cows, stage of lactation, and milk yield. Alkaline phosphatase is inactivated at a higher temperature than the usual temperature of pasteurization. Decreased activity of alkaline phosphatase can therefore be used to determine that milk was pasteurized properly and that the pasteurized milk is not contaminated with raw milk. Alkaline phosphatase is able to cleave phenolphthalein monophosphate releasing phenolphthalein. Hardy Diagnostics Alkaline Phosphatase Kit is based on the Rutgers method, which is a stable and sensitive method that provides an easy to interpret color read out within 30 minutes.

Specifications

Specifications

  • Description:
    Alkaline phosphatase kit, 125 tests/kit
  • Volume:
    3×7 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.