ONPG, Rapid Test Broth, Hardy Diagnostics
Catalog # 75845-730
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:PC tube
- Description:ONPG, Rapid Test Broth, HDx
- Size:13 x 100 mm
- Cat. no.:75845-730
- Volume:0.5 ml
- Supplier No.:R92
Specifications
About this item
For the rapid determination of beta-Galactosidase activity of gram negative bacilli.