Oxoid Novobiocin Supplement, Hardy Diagnostics
Catalog # CA89423-158
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Description:Novobiocin Supplement
- Size:10 x 10 mg
- Cat. no.:CA89423-158
- Supplier No.:SR181E
Specifications
About this item
A selective supplement for the addition to culture media to enhance inhibition of Gram positive organisms.
A selective supplement for the addition to culture media to enhance inhibition of Gram positive organisms.