To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:LB Broth, Lennox, HDx
- Size:Pack of 6
- Cat. no.:CA89501-486
- Volume:500 mL
- Supplier No.:C7659
Specifications
About this item
recommended for maintaining and cultivating recombinant strains of Escherichia coli in molecular biology procedures.
Hardy Diagnostics CulGenex™ LB Broth, Lennox is recommended for maintaining and cultivating recombinant strains of Escherichia coli for molecular biology procedures.
Hardy Diagnostics CulGenex™ Dehydrated Culture Media is formulated to meet or exceed the highest standards. Each lot of CulGenex™ media is meticulously tested for performance and reproducibility. Over 30 years of formulating and media manufacturing experience.
Container sizes available:
2L mylar pouch – pre-measured for a 2 liter batch, no weighing needed!
500ml mylar pouch - pre-measured for a 500ml batch, 6 pouches per pack
500gm bottle - wide mouth, easy grip bottle, comes with a free scoop!
2kg and 10kg bucket - with convenient screw lid