Order Entry
Canada
ContactUsLinkComponent
Hemin and Vitamin K Solution, Hardy Diagnostics
Hemin and Vitamin K Solution, Hardy Diagnostics
Catalog # 75803-006
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Hemin and Vitamin K Solution, Hardy Diagnostics
Catalog # 75803-006
Supplier:  Hardy Diagnostics
Supplier Number:  Z237
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    Hemin & Vitamin K, HDx
  • Size:
    16 x 125 mm
  • Cat. no.:
    75803-006
  • Volume:
    10 ml
  • Supplier No.:
    Z237

Specifications

About this item

A growth supplement for fastidious anaerobic bacteria.

(Hemin, 0.25mg/ml and Vitamin K, 0.050mg/ml) A growth supplement for fastidious anaerobic bacteria.

Ordering information: Minimum order of 5 packs