Order Entry
Canada
Orders LinkContactUsLinkComponent
Hayflick Medium Agar

Hayflick Medium Agar

Hayflick Medium Agar
Supplier:  Hardy Diagnostics
Certificates
Hayflick Medium Agar
Hayflick Medium Agar

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recommended for use in the cultivation and isolation of Mycoplasma species.

Ordering information: Minimum order of 10 packs

Documents

Product Family Options

Product Information

  • Size
    Volume
    Format
    Application
    Dimensions
    Minimum Order Quantity
    Availability
    Price
  • 10124-546
    Hayflick medium agar
    Hayflick medium agar
    10/pack
    11 ml
    Plate, small
    Clinical
    15×60 mm
    10
    Log in for availability
    Log in for availability
    Each
    Each - $143.00
    $143.00
    Each
       $143.00
    Minimum order quantity is 10

    Supplier Number: G385
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • CA10124-546
    Hayflick medium agar
    Hayflick medium agar
    10/pack
    11 ml
    Plate, small
    Clinical
    15×60 mm
    10
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $179.00
    $179.00
    Pack of 10
       $179.00
    Minimum order quantity is 10

    Supplier Number: G385
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.