ContactUsLinkComponent
HardyDisks™ AST Ceftaroline, CPT-30, Hardy Diagnostics
HardyDisks™ AST Ceftaroline, CPT-30, Hardy Diagnostics
Catalog # 75802-976
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
HardyDisks™ AST Ceftaroline, CPT-30, Hardy Diagnostics
Catalog # 75802-976
Supplier:  Hardy Diagnostics
Supplier Number:  Z9391
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Ceftaroline, CPT-30, HDx
  • Size:
    1 x 50 disks
  • Cat. no.:
    75802-976
  • Supplier No.:
    Z9391
Specifications
About this item
Impregnated paper disks used for Antimicrobial Susceptibility Testing (AST)
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.