Order Entry
ContactUsLinkComponent
36 results for Centrifuges

You searched for: Microcentrifuges

Microcentrifuges

Compact microcentrifuges isolate DNA or simply separate micro volume mixtures. Load microtubes and standard or dell-welled plates in the instruments easily. Adaptable to any industrial or laboratory setting, non-refrigerated or refrigerated models come with multiple mode options for the high-speed and powerful rotors. Continuous or pulse spinning options move samples fast and consistently while included timers alert with sample preparation is complete. Microcentrifuges may come with or without the protective lid.

Sort by
VWR® High Speed Microcentrifuge

VWR® High Speed Microcentrifuge

Supplier: VWR International

The compact, affordable VWR® High Speed Microcentrifuge is capable of high speed performance up to 15500 rpm/16100×g.

Expand 1 Items
Loading...
VWR® Mini Centrifuges

VWR® Mini Centrifuges

Supplier: VWR International

The VWR® Mini Centrifuge is designed for quick spin-downs of microcentrifuge tubes and PCR strips and tubes.

Expand 1 Items
Loading...
VWR® Compact Micro Centrifuges

VWR® Compact Micro Centrifuges

Supplier: VWR International

These compact microcentrifuges are ideal for DNA, RNA, and temperature-sensitive samples.

Expand 9 Items
Loading...
VWR® Plate Centrifuge

VWR® Plate Centrifuge

Supplier: VWR International

Spins at 2500 rpm/ 850xg, to quickly spin down droplets and condensation in PCR plates.

Expand 1 Items
Loading...
VWR® Micro Star 21 and 21R Microcentrifuges

VWR® Micro Star 21 and 21R Microcentrifuges

Supplier: VWR International

VWR® Micro Star 21 ventilated and 21R refrigerated microcentrifuge with 24×1.5/2.0 ml rotor with ClickSeal biocontainment lid offer exceptional acceleration of up to 21000 xg in as little as 12 seconds.

Expand 2 Items
Loading...
Hettich MIKRO 185 Microliter Centrifuges, Hettich Lab Technology

Hettich MIKRO 185 Microliter Centrifuges, Hettich Lab Technology

Supplier: HETTICH INSTRUMENTS

Hettich Lab Technology's MIKRO 185 tabletop microliter centrifuge delivers application versatility and fast results in an elegant, compact frame.

Expand 3 Items
Loading...
Microcentrifuges, 230 V, 5418/5418R

Microcentrifuges, 230 V, 5418/5418R

Supplier: Eppendorf

Ultra-compact, 18-place microcentrifuges are quiet and user-friendly

Expand 1 Items
Loading...
Stuart® 8500 rpm SCF4 Microcentrifuge, Cole Parmer

Stuart® 8500 rpm SCF4 Microcentrifuge, Cole Parmer

Supplier: Argos Technologies

The Stuart 8500 microcentrifuge provides high speed centrifugation with enhanced safety features.

Expand 1 Items
Loading...

Microcentrifuge, 5425, GLP, Eppendorf

Supplier: Eppendorf

Microcentrifuge 5425 plus the 1.5/2 ml kit rotor is the optimal choice for spin-column-based nucleic acid purification kits containing multiple centrifugation steps with frequent vessel changes, as used in target antigen production, sequencing sample preparation and antibody humanization.

Expand 12 Items
Loading...
Microcentrifuges, 230 V, 5430 and 5430R

Microcentrifuges, 230 V, 5430 and 5430R

Supplier: Eppendorf

The unique cross-over microcentrifuges 5430 and 5430 R combine the features of a microcentrifuge (small footprint) and multipurpose centrifuge (versatility) in one instrument.

Expand 5 Items
Loading...

Eppendorf 5427R Microcentrifuges

Supplier: Eppendorf

Labs demanding high sample throughput will benefit from the 5427R, featuring a double-row 48-place rotor to increase efficiency and improve running time.

Expand 1 Items
Loading...
Hettich MIKRO 220 Microliter Centrifuges, Hettich Lab Technology

Hettich MIKRO 220 Microliter Centrifuges, Hettich Lab Technology

Supplier: HETTICH INSTRUMENTS

The Hettich Lab Technology MIKRO 220 microliter centrifuge series offers large capacities and high-performance for high-throughput laboratories.

Expand 3 Items
Loading...
Eppendorf® Microcentrifuges, Models 5430 and 5430R

Eppendorf® Microcentrifuges, Models 5430 and 5430R

Supplier: Eppendorf

The unique cross-over microcentrifuges 5430 and 5430 R combine the features of a microcentrifuge (small footprint) and multipurpose centrifuge (versatility) in one instrument.

Expand 17 Items
Loading...
Eppendorf® MiniSpin® Personal Microcentrifuges

Eppendorf® MiniSpin® Personal Microcentrifuges

Supplier: Eppendorf

Microcentrifuges are compact and easy to use, yet powerful enough to enable each workstation to be equipped with a “personal” centrifuge for a wide range of molecular biology separations and quick spins

Expand 2 Items
Loading...
Microfuge® 16 Microcentrifuge, Beckman Coulter®

Microfuge® 16 Microcentrifuge, Beckman Coulter®

Supplier: Beckman Coulter

This non-refrigerated, cold-room compatible, high-performance microcentrifuge is ideal for fast pelleting or isolation of DNA, RNA, proteins, and viruses.

Expand 1 Items
Loading...
Corning® LSE™ Mini Microcentrifuge, Corning

Corning® LSE™ Mini Microcentrifuge, Corning

Supplier: Corning

The Corning® LSE™ mini microcentrifuge is a personal bench top instrument that operates at a fixed speed of 6000 min⁻¹ (2000×g) and was designed for quick spin downs of micro-samples.

Expand 1 Items
Loading...
mySPIN™ Mini Centrifuges Series, Thermo Scientific

mySPIN™ Mini Centrifuges Series, Thermo Scientific

Supplier: THERMO FISHER SCIENTIFIC MX

Bring increased efficiency and user comfort to your lab with mySPIN™ 6 and 12 mini centrifuges.

Expand 2 Items
Loading...
Eppendorf® Microcentrifuges, 5418/5418R

Eppendorf® Microcentrifuges, 5418/5418R

Supplier: Eppendorf

Ultra-compact, 18-place microcentrifuges are quiet and user-friendly

Expand 1 Items
Loading...
Hettich MIKRO 200 Microliter Centrifuges, Hettich Lab Technology

Hettich MIKRO 200 Microliter Centrifuges, Hettich Lab Technology

Supplier: HETTICH INSTRUMENTS

The Hettich MIKRO 200 microliter centrifuge series is amongst the highest performing in its class.

Expand 2 Items
Loading...
Eppendorf 5427R Centrifuges with Hydrocarbon Cooling

Eppendorf 5427R Centrifuges with Hydrocarbon Cooling

Supplier: Eppendorf

Eppendorf Centrifuge 5427 R supports your greener laboratory with hydrocarbon cooling. It is the perfect solution for laboratories with a variety of molecular biology applications or those in need of high-throughput solutions for 1.5 to 2 ml tubes - all combined in a microcentrifuge with the highest throughput/footprint-ratio within the Eppendorf microcentrifuge range.

Expand 3 Items
Loading...
Eppendorf 5427R Centrifuges with Hydrocarbon Cooling, 230 V

Eppendorf 5427R Centrifuges with Hydrocarbon Cooling, 230 V

Supplier: Eppendorf

Eppendorf Centrifuge 5427 R supports your greener laboratory with hydrocarbon cooling. It is the perfect solution for laboratories with a variety of molecular biology applications or those in need of high-throughput solutions for 1.5 to 2 ml tubes - all combined in a microcentrifuge with the highest throughput/footprint-ratio within the Eppendorf microcentrifuge range.

Expand 1 Items
Loading...
Microfuge® 20 Series IVD Microcentrifuges, Beckman Coulter®

Microfuge® 20 Series IVD Microcentrifuges, Beckman Coulter®

Supplier: Beckman Coulter

Fast and versatile, the high-performance Microfuge® 20 Series are precisely designed to meet a wide variety of application needs.

Expand 6 Items
Loading...
Mini Centrifuges, D1008, Dlab Scientific

Mini Centrifuges, D1008, Dlab Scientific

Supplier: DLAB SCIENTIFIC INC.

The D1008 Mini centrifuge is ideal for microfiltration and quick spin-downs.

Expand 1 Items
Loading...
Labnet Prism R Refrigerated Microcentrifuge

Labnet Prism R Refrigerated Microcentrifuge

Supplier: LABNET INTERNATIONAL SE

This high performance, refrigerated centrifuge can cool to 4°C in just eight minutes.

Expand 1 Items
Loading...
Ohaus® Frontier™ 5000 Series Mini Centrifuge

Ohaus® Frontier™ 5000 Series Mini Centrifuge

Supplier: OHAUS CORP.

The Ohaus Frontier 5306 is a small but powerful minicentrifuge for simple and quick liquid separation.

Expand 1 Items
Loading...
Cole-Parmer® CF-200 Series Stuart Programmable Microcentrifuge, Antylia Scientific

Cole-Parmer® CF-200 Series Stuart Programmable Microcentrifuge, Antylia Scientific

Supplier: Argos Technologies

Compact, powerful, and programmable.

Expand 1 Items
Loading...
Corning® LSE™ High Speed Microcentrifuge, Corning

Corning® LSE™ High Speed Microcentrifuge, Corning

Supplier: Corning

The Corning® LSE High Speed Microcentrifuge features an easy-to-use, digital control interface and high-speed performance for quick nucleic acid and protein separations

Expand 1 Items
Loading...
VWR® High Speed Microcentrifuge, 220 V

VWR® High Speed Microcentrifuge, 220 V

Supplier: BENCHMARK SCIENTIFIC

The compact, affordable VWR® High Speed Microcentrifuge is capable of high speed performance up to 15500 rpm/16100×g.

Expand 1 Items
Loading...
SPROUT® Plus Mini Centrifuges

SPROUT® Plus Mini Centrifuges

Supplier: Heathrow Scientific

Mini but mighty - ideal for small runs.

Expand 2 Items
Loading...
HemataSTAT® II Centrifuge, Separation Technology

HemataSTAT® II Centrifuge, Separation Technology

Supplier: Separation Technology

This microhematrocrit system provides a quantitative measure of the amount of red blood cells in relation to the amount of plasma

Expand 1 Items
Loading...
Sort by
Recommended for You