ContactUsLinkComponent
Buffered Sodium Chloride-Peptone Solution, USP
Buffered Sodium Chloride-Peptone Solution, USP
Catalog # CA89407-430
CAS Number:  
undefined
Buffered Sodium Chloride-Peptone Solution, USP
Catalog # CA89407-430
Supplier Number:  U255
CAS Number:  
Specifications
  • Pack type:
    Jar, wide mouth
  • Description:
    Buffered sodium chloride-peptone solution, USP
  • Cat. no.:
    CA89407-430
  • Volume:
    90 ml
  • Supplier No.:
    U255
Specifications
About this item
For the preparation of isotonic suspensions and dilutions of microorganisms.
For the preparation of isotonic suspensions and dilutions of microorganisms.