Buffer No. 4
Buffer no.4, USP, 500 ml, PC bottle, 500 ml
Catalog # CA89407-432
Supplier: HARDY DIAGNOSTICS CA
Supplier #: U307
Jump to:
Product Details & Documents
For use in antibiotic potency assay.
For use in antibiotic potency assay.
Ordering information: Minimum order of 5 packs of 10
Documents
Certificates
Search by Lot NumberHardy Diagnostics Pharmaceutical Microbiology Products
Product Literature
Specifications
Specifications
- Pack type:PC bottle
- Description:Buffer no.4, USP
- Size:500 ml
- Volume:500 ml
- Cat. No.:CA89407-432
- Storage temperature:–2...+30 °C



