Order Entry
Canada
ContactUsLinkComponent
Bile Spot Test, Hardy Diagnostics
Bile Spot Test, Hardy Diagnostics
Catalog # CA10790-366
CAS Number:  
undefined
Bile Spot Test, Hardy Diagnostics
Catalog # CA10790-366
Supplier Number:  Z61
CAS Number:  

Specifications

  • Description:
    Bile Spot Test, HDx, Deoxych 10%
  • Cat. no.:
    CA10790-366
  • Volume:
    15 ml
  • Supplier No.:
    Z61

Specifications

About this item

Bile Spot Test, Deoxycholate 10% is used to identify S

  • Colonies will autolyze in the presence of bile salts

Sodium desoxycholate 10%, use to identify S. pneumoniae, colonies will autolyze in the presence of bile salts. Store in refrigerator, warm before use.
Recommended for use in presumptively differentiating bile soluble Streptococcus pneumoniae from bile insoluble alpha-hemolytic Streptococcus spp. The bile solubility test may be performed using a cell suspension or by applying the Bile Spot Reagent directly to the colony.

pneumoniae.