ALP Urea, Modified, Hardy Diagnostics
Catalog # 75803-052
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Tube
- Description:Modified ALP Urea, HDx
- Size:13 x 100 mm
- Minimum Order Quantity:5
- Cat. no.:75803-052
- Volume:3 ml
- Supplier No.:Y70
Specifications
About this item
ALP Urea, Modified
(Aerobic Low Peptone) For the identification of non-fermentative bacilli. See technical insert.
Ordering information: Minimum Order - 5 packs