Acetate Differential Slant, Hardy Diagnostics
Catalog # 75803-034
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Tube
- Description:Acetate Differential Slant
- Size:16 x 100 mm
- Minimum Order Quantity:5
- Cat. no.:75803-034
- Supplier No.:L15
Specifications
About this item
Acetate Differential Slant For the differentiation of E.coli and Shigella spp.
Ordering information: Minimum Order - 5 packs