Order Entry
Canada
ContactUsLinkComponent
 
IdeZ Protease (IgG-specific)
 
IdeZ Protease (IgG-specific)

 

  • Source:
    E. coli
  • Species:
    Rabbit
    Monkey
    Sheep
    Human
  • Storage conditions:
    -20 °C
  • Enzyme name:
    IdeZ Protease (IgG-Specific)
  • Concentration:
    80,000 units/ml
  • Tested applications:
    Proteomics

 

Frequently Bought Together