Order Entry
Canada
ContactUsLinkComponent
Anti-CD20 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 2H7]
Anti-CD20 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 2H7]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD20 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: 2H7]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 75834-358
  • 75834-360
  • Antigen symbol:
  • CD20
  • CD20
  • Antigen name:
  • CD20
  • CD20
  • Antigen synonyms:
  • Bp35|B1|Leu-16
  • Bp35|B1|Leu-16
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Clone:
  • 2H7
  • 2H7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 931
  • 931
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light

Specifications

Related Information

Frequently Bought Together