Order Entry
Canada
ContactUsLinkComponent
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody (APC-Cyanine7) [clone: M5/114.15.2]
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody (APC-Cyanine7) [clone: M5/114.15.2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody (APC-Cyanine7) [clone: M5/114.15.2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 75834-356
  • 75834-354
  • Antigen symbol:
  • MHC Class II (I-A/I-E)
  • MHC Class II (I-A/I-E)
  • Antigen name:
  • MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek
  • MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek
  • Antigen synonyms:
  • MHC II|IA/IE|IA|IE|I-A/E
  • MHC II|IA/IE|IA|IE|I-A/E
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • M5/114.15.2
  • M5/114.15.2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 14961/14969
  • 14961/14969
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light

Specifications

Related Information

Frequently Bought Together