Order Entry
Canada
ContactUsLinkComponent
Anti-CD62L Rat Monoclonal Antibody (APC-Cyanine7) [clone: MEL-14]
Anti-CD62L Rat Monoclonal Antibody (APC-Cyanine7) [clone: MEL-14]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD62L Rat Monoclonal Antibody (APC-Cyanine7) [clone: MEL-14]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 75834-290
  • 75834-292
  • Antigen symbol:
  • CD62L
  • CD62L
  • Antigen name:
  • CD62L, L-Selectin
  • CD62L, L-Selectin
  • Antigen synonyms:
  • Ly-22|LECAM-1
  • Ly-22|LECAM-1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • MEL-14
  • MEL-14
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 20343
  • 20343
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light

Specifications

Related Information

Frequently Bought Together