Order Entry
Canada
Orders LinkContactUsLinkComponent
 

BBL™ Phosphate Buffer, pH 7.2, BD

BBL™ Phosphate Buffer, pH 7.2, 500 g
Catalog # CA90000-224
Supplier:   BD
 :  211544
BBL™ Phosphate Buffer, pH 7.2, BD
Catalog # CA90000-224
Supplier:  BD
CAS Number:  
BBL™ Phosphate Buffer, pH 7.2, BD

BBL™ Phosphate Buffer, pH 7.2, 500 g

CAS Number:  
Price
$174.00
Log in for availability
Each (500g)
Product Details & Documents

Phosphate Buffer, pH 7.2 is used for the preparation of dilution blanks for use in the examination of waters, dairy products, foods, and other materials.

  • Sodium carbonate is a pH regulator
  • Prepared appearance: Colorless to light yellow and clear to trace hazy

Phosphate buffer is used in the preparation of dilution blanks for use in microbiological testing rather than unbuffered water in order to standardize this potential variable due to the wide variation in the pH of puriifed water from multiple sources.

Specifications

Specifications

  • Description:
    BBL™ Phosphate Buffer, pH 7.2
  • Size:
    500 g

Specifications