Order Entry
Canada
ContactUsLinkComponent
Difco™ Pantothenate Medium AOAC, BD Diagnostics
Difco™ Pantothenate Medium AOAC, BD Diagnostics
Catalog # CA90004-034
Supplier:  BD
CAS Number:  
Difco™ Pantothenate Medium AOAC, BD Diagnostics
Catalog # CA90004-034
Supplier:  BD
Supplier Number:  281610
CAS Number:  

Specifications

  • Description:
    Difco™ Pantothenate Medium AOAC
  • Size:
    100 g
  • Cat. no.:
    CA90004-034
  • Supplier No.:
    281610

Specifications

About this item

Pantothenate Medium AOAC is used for determining the concentration of pantothenic acid and pantothenate by the microbiological assay technique.

  • Does not contain pantothenic acid or pantothenate
  • Contains all other essential nutrients
  • Calcium pantothenate stimulates a growth response
  • Does not contain polysorbate 80

Pantothenate Medium AOAC is prepared for use in the microbiological assay of pantothenic acid and pantothenate according to the procedures of Calcium Pantothenate Assay in the USP1 and Pantothenate Acid Assay in the Official Methods of Analysis of AOAC International (AOAC).

Pantothenate Medium AOAC is a pantothenic acid/pantothenate-free dehydrated medium containing all other nutrients and vitamins essential for the cultivation of Lactobacillus plantarum ATCC™ 8014.