ContactUsLinkComponent
General Purpose Tray, with Econo Floor Grade Instruments, Sklar
General Purpose Tray, with Econo Floor Grade Instruments, Sklar
Catalog # 75817-998
Supplier:  Sklar
undefined
General Purpose Tray, with Econo Floor Grade Instruments, Sklar
Catalog # 75817-998
Supplier:  Sklar
Supplier Number:  96-1700
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    General Purpose Tray, with Econo Floor Grade Instruments
  • Cat. no.:
    75817-998
  • Packaging:
    25/Case
Specifications
About this item
General Purpose Trays are industry standard
Each tray includes the following instruments and prep materials: Curved Hemostat, Tissue Forceps, OR Scissors, S/B, PVP Swabstick, Alcohol Swabstick, (2) Cotton Tip Applicators, (2) Gauze Sponges (4x4", 12-ply), Safety Pin #2, and Shallow Procedure Tray.