BioWhittaker® DMEM Cell Culture Media, Lonza
Supplier: Custom Comfort
Certificates
About this item
DMEM is a modification of Basal Medium Eagle (BME) that contains four times the concentrations of amino acids and vitamins.
- Suitable for most types of cells
- Basal Medium Eagle modification
- Four times the vitamin and amino acid concentration
DMEM is a modification of Basal Medium Eagle (BME) that contains four times the concentrations of amino acids and vitamins. The formulation first contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. DMEM media is suitable for most types of cells, including human, monkey, hamster, rat, mouse, chicken and fish cells.
All BioWhittaker™ Cell Culture Media products are manufactured in accordance with cGMP regulations. The formulations used for standard liquid media products are those recommended by the Tissue Culture Association.
Specifications
- Storage temperature:2...8 °C
Specifications
Related Information
Certificates
Search by Lot Number12001-576
Datasheet
Lonza BioWhittaker Classical Media
Product Literature
Frequently Bought Together