ContactUsLinkComponent
Concepts of Molarity
Concepts of Molarity
Catalog # 470220-224
Supplier:  Ward's Science
undefined
Concepts of Molarity
Catalog # 470220-224
Supplier:  Ward's Science
Supplier Number:  IS2519
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Molarity Lab Investigation
  • No. of Lab Groups:
    15
  • Cat. no.:
    470220-224
  • Supplier No.:
    IS2519
Specifications
About this item
Designed to Introduce Students to the Concept of Molarity.
  • Practice proper lab techniques
  • Visually determine the unknowns
Students will first try to make a solution with a specific molarity demonstrating the importance of good laboratory technique. In the second set of experiments students will perform a titration on a known acid solution before using the concepts they have learned to identify the concentrations in three unknown solutions.
Ordering information: Kit includes Hydrochloric Acid, Phenolphthalein Solution, Sodium Hydroxide, Sucrose, Unknown Hydrochloric Acids #1-#3, Teacher's Manual, and Student Study Guide Copymasters. Materials needed but not included are (15) 100 mL Beakers, (15) 200+ mL Beakers,(15-30) Burettes, (15) Burette Funnels, (15) Burette Clamps, (15) Burette Stands, Deionized Water, Graduated Cylinder, Permanent Marker, and Stirring Rod.