Aldehyde Dehydrogenase
Catalog # CA80108-488
Supplier: MilliporeSigma
CAS Number:
Specifications
- CAS No.:9028-88-0
- Size:100 units
- Storage conditions:2...8 °C
- Form:white lyophilized powder
- Enzyme name:Aldehyde dehydrogenase
- Cat. no.:CA80108-488
- Supplier No.:126925-100U
Specifications
About this item
Aldehyde dehydrogenase from yeast catalyzes the oxidation of acetaldehyde to acetic acid.