ContactUsLinkComponent
BBL™ Thioglycollate Medium, Brewer Modified, BD Life Sciences
  CA90000-294
 :  BD
 :  
BBL™ Thioglycollate Medium, Brewer Modified, BD Life Sciences
  CA90000-294
 :  BD
 :  211716
 :  
  • Description:
    BBL™ Thioglycollate Medium, Brewer Modified
  • Size:
    500 g
  • Cat. no.:
    CA90000-294
  • Supplier No.:
    211716
Thioglycollate Medium, Brewer Modified is used for testing sterility of biological products and for the isolation of aerobic and anaerobic organisms.
  • Modified for sterility testing of products containing mercury preservatives
  • Peptones, beef extract, and yeast extract provide essential nutrients
  • Suitable for luxuriant growth of fastidious microorganisms
  • Very small amount of agar helps in maintaining anaerobic condition
The modified medium contains more thioglycollate and is recommended for sterility testing procedure.