ContactUsLinkComponent
Difco™ Tetrathionate (TT) Broth Base, Hajna, BD Biosciences
Difco™ Tetrathionate (TT) Broth Base, Hajna, BD Biosciences
Catalog # CA90003-424
Supplier:  BD
CAS Number:  
Difco™ Tetrathionate (TT) Broth Base, Hajna, BD Biosciences
Catalog # CA90003-424
Supplier:  BD
Supplier Number:  249120
CAS Number:  
Specifications
  • Description:
    Difco™ Tetrathionate (TT) Broth Base, Hajna
  • Size:
    500 g
  • Cat. no.:
    CA90003-424
  • Supplier No.:
    249120
Specifications
About this item
TT Broth Base, Hajna is used for enriching Salmonella from food and dairy products prior to isolation procedures.
  • Peptone provides nitrogen and amino acids
  • Yeast extract supplies growth factors and vitamins
  • Dextrose and mannitol are fermentable carbohydrates
  • Calcium carbonate is a neutralizer that absorbs toxic metabolites
Organisms containing the enzyme tetrathionate reductase will proliferate in this medium.