Difco™ Supplement B with Reconstituting Fluid, BD Biosciences
Catalog # CA90001-934
Supplier: BD
CAS Number:
Specifications
- Description:Difco™ Supplement B with Reconstituting Fluid
- Size:10 mL
- Cat. no.:CA90001-934
- Supplier No.:227610
Specifications
About this item
Supplement B with Reconstituting Fluid is used for supplementing media to culture fastidious microorganisms, particularly Neisseria gonorrhoeae and Haemophilus influenzae.
- Contains yeast concentrate, glutamine, coenzyme, cocarboxylase, hematin, and growth factors
- For use in augmenting the preparation of chocolate agar
Supplement B is a yeast concentrate for use in supplementing media for growth of microorganisms with exacting nutritional requirements.