Difco™ Select APS™ Super Broth Base, BD Diagnostics
: BD
Select APS™ Super Broth is a molecular genetics medium used to grow Escherichia coli to a high cell density.
- Molecular genetics medium
- Alternative to classical animal-based media
- Allows maintenance and propogation of E. coli
- Soy hydrolysate provides nitrogen and carbon
- Glycerol is added as a carbon and energy source
Select APS™ media are manufactured from animal-free ingredients in order to minimize the risk of bovine spongiform encephalopathy in culture media containing animal, and especially bovine, materials.
Select APS™ Super Broth is a molecular genetics medium used to grow Escherichia coli to a high cell density.
- Molecular genetics medium
- Alternative to classical animal-based media
- Allows maintenance and propogation of E. coli
- Soy hydrolysate provides nitrogen and carbon
- Glycerol is added as a carbon and energy source
Select APS™ media are manufactured from animal-free ingredients in order to minimize the risk of bovine spongiform encephalopathy in culture media containing animal, and especially bovine, materials.
Product Family Options
- DescriptionSize
- Difco™ Select APS™ Super Broth Base10 kgEach (10kg)$2,810.00: 212486
- : 212485