Sabouraud Dextrose Broth, BD
Catalog # CA95038-630
Supplier: BD
CAS Number:
Specifications
- Description:Sabouraud Dextrose Broth
- Size:100 mL
- Cat. no.:CA95038-630
- Supplier No.:215193
Specifications
About this item
Sabouraud Dextrose Broth is used for the cultivation of yeasts, molds, and aciduric microorganisms.
- Peptone provides nitrogen, vitamins, minerals, amino acids, and growth factors
- Low pH promotes the growth of fungi
- Dehydrated appearance: Cream to yellow, free-flowing
The addition of antimicrobics further increases the selectivity of the medium.