Orders LinkContactUsLinkComponent
R2A Agar, BD
 :   BD
R2A Agar, BD
 :  BD
R2A Agar, BD
R2A Agar is used for enumerating heterotrophic organisms in treated potable water.
  • Dehydrated appearance: Light beige, flree-flowing
  • Prepared appearance: Light amber, slightly opalescent
  • Improves recovery of chlorine-tolerant bacteria
A low-nutrient medium, R2A Agar is used in combination with a lower incubation temperature and longer incubation time to stimulate the growth of stressed and chlorine-tolerant bacteria.
Product Family Options