m Staphylococcus Broth, BD Life Sciences
m Staphylococcus Broth, 500 g
Supplier: BD
Supplier #: 264920
Jump to:
Product Details & Documents
m Staphylococcus Broth is used for isolating staphylococci by the membrane filtration technique.
- Staphylococci are indicators of recreational water quality
- Used in a multiple-tube procedures
- For monitoring swimming pool water
- For the identification of S. aureus
- Dehydrated appearance: Light beige, free-flowing
Observe membranes for growth and pigment production. Test for mannitol fermentation by adding a drop of bromthymol blue to the site from which a colony is removed; a yellow color indicates mannitol fermentation.
Documents
Specifications
Specifications
- Description:m Staphylococcus Broth
- Size:500 g