Orders LinkContactUsLinkComponent
Biox Transfer Pipettes
Supplier:   TWD Scientific
Biox Transfer Pipettes
Supplier:  TWD Scientific
Certificates
Biox Transfer Pipettes
Biox Transfer Pipettes
4 Options Available Below
Product Details & Documents
Transfer Pipettes are perfect for quick and safe liquid transfer.
  • Disposable
  • Polyethylene construction
  • Uniform drop size
  • 100% inert for accurate sampling
Polyethylene pipettes are a great alternative to pasteur pipettes or rubber bulbs.
Product Family Options
Product Information
  • Length
    Sterility
    Graduations
    Packed
    Availability
    Price
  • 160 mm
    Sterile
    To 1 mL
    Individually Wrapped
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $800.50
    $800.50
    Case of 2,000
       $800.50
    Supplier Number: BX30-0135A1-1
  • 160 mm
    Non sterile
    To 1 mL
    Bulk
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $324.37
    $324.37
    Case of 2,000
       $324.37
    Supplier Number: BX30-0135-1
  • 160 mm
    Sterile
    To 3 mL
    Individually Wrapped
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $825.90
    $825.90
    Case of 2,000
       $825.90
    Supplier Number: BX30-0138A1-1
  • 160 mm
    Non sterile
    To 3 mL
    Bulk
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $357.16
    $357.16
    Case of 2,000
       $357.16
    Supplier Number: BX30-0138-1