Orders LinkContactUsLinkComponent
PCR Tubes
PCR Tubes
Supplier:   TWD Scientific
PCR Tubes
Supplier:  TWD Scientific
Certificates
PCR Tubes
1 Options Available Below
Product Details & Documents
PCR tubes are ideally suited for real-time PCR applications.
  • Prime virgin PP construction
  • Temperature: stable from –20 to 100 °C (–4 to 212 °F)
  • Tubes withstand centrifugation of 6000 RCF
  • Autoclavable in open state
  • Ultra-thin wall tubes
Rnase-, Dnase-, and endotoxin free.
Product Family Options
Product Information
  • Color
    Capacity
    Cap type
    Availability
    Price
  • 75799-968
    Individual Tubes
    Individual Tubes
    Clear
    0.2 ml
    Dome-shaped cap
    Discontinued
    Supplier Number: -
  • 75799-966
    Individual Tubes
    Individual Tubes
    Clear
    0.2 ml
    Flat top
    Discontinued
    Supplier Number: -
  • 75799-972
    8-Strip Tubes
    8-Strip Tubes
    Clear
    0.2 ml
    Dome-shaped cap
    Log in for availability
    Log in for availability
    Case of 1,250
    Case of 1,250 - $1,176.81
    $1,176.81
    Case of 1,250
       $1,176.81
    Supplier Number: BX60-0008