Orders LinkContactUsLinkComponent
Biox Cardboard Freezer Boxes
Biox Cardboard Freezer Boxes
Supplier:   TWD Scientific
Biox Cardboard Freezer Boxes
Supplier:  TWD Scientific
Certificates
Biox Cardboard Freezer Boxes
Biox Cardboard Freezer Boxes
4 Options Available Below
Product Details & Documents
Premium cardboard freezer boxes with a water-resistant coating.
  • Chemically resistant to alcohols and mild organic solvents
  • Temperature range: –196 to 121°C (–320 to 250 °F)
  • Accommodates 0.5 ml, 1.5 ml, or 2.0 ml microtubes
  • Suitable for liquid nitrogen storage
Boxes feature a water-resistant coating for improved durability.
Product Family Options
Product Information
  • No. of Holes
    Height
    Availability
    Price
  • 81
    5 cm (2")
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $603.30
    $603.30
    Case of 100
       $603.30
    Supplier Number: BX90-2281
  • 100
    5 cm (2")
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $553.75
    $553.75
    Case of 100
       $553.75
    Supplier Number: BX90-2200
  • 81
    7.6 cm (3")
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $638.35
    $638.35
    Case of 100
       $638.35
    Supplier Number: BX90-2381
  • 100
    7.6 cm (3")
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $620.95
    $620.95
    Case of 100
       $620.95
    Supplier Number: BX90-2300