Order Entry
Canada
Orders LinkContactUsLinkComponent
BBL™ Gram Basic Fuchsin, BD

BBL™ Gram Basic Fuchsin, BD

BBL™ Gram Basic Fuchsin, 3.8 L
Catalog # CA90000-716
Supplier:   BD
Supplier #:  212545
BBL™ Gram Basic Fuchsin, BD
Catalog # CA90000-716
Supplier:  BD
CAS Number:  
BBL™ Gram Basic Fuchsin, BD

BBL™ Gram Basic Fuchsin, 3.8 L

CAS Number:  
Price
$1,300.00
Log in for availability
Each (4l)
Product Details & Documents

Gram stain reagents are used to stain microorganisms from cultures or specimens by the differential Gram method.

  • Differential Gram method
  • Stains more intensely than safranin

Basic fuchsin stains many Gram-negative bacteria more intensely than safranin, making them easier to see.

Specifications

Specifications

  • Description:
    BBL™ Gram Basic Fuchsin
  • Size:
    3.8 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.