Order Entry
Canada
ContactUsLinkComponent
Ultra Cyano HPLC Prep Columns, Restek
Ultra Cyano HPLC Prep Columns, Restek
Catalog # 10059-668
Supplier:  Restek
undefined
Ultra Cyano HPLC Prep Columns, Restek
Catalog # 10059-668
Supplier:  Restek
Supplier Number:  9106577

Specifications

  • Column Format:
    Semi-preparative
  • Ø int.:
    10.00 mm
  • Particle Size:
    5 µm
  • Pore size:
    100 Å
  • Description:
    HPLC Prep Column
  • Length:
    250 mm
  • Endcapped:
    Yes
  • pH range:
    2.5 to 8
  • Temperature limit:
    80°C
  • Carbon load:
    Carbon Load: 8%
  • Cat. no.:
    10059-668
  • Packing Material:
    Cyanopropylsilane
  • Phase:
    Ultra Cyano
  • Supplier No.:
    9106577

Specifications

About this item

Restek makes it easy to scale up your separations.

  • Easy scale-up from Restek analytical columns
  • Popular bonded phases

We offer a wide range of semipreparative and preparative-scale columns packed with many of our popular Ultra, Allure®, Pinnacle® DB, and Viva bonded phases on a 5 μm spherical silica.

The Restek® Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired. It can be used in normal-phase, reversed-phase (best with mobile phase pH between 5 and 7), and HILIC modes. It is an excellent choice for the analysis of protonated bases.

Other stationary phases are also available.

Ordering information: We strongly recommend ordering a semi-prep or prep column only after evaluating the desired separation on an equivalent analytical-scale column. Because we cannot reuse a column or the silica it contains once it has left our facility, we cannot accept returns of large-scale columns.