MacConkey Agar w/o Crystal Violet, BD
Supplier: BD
Certificates
About this item
MacConkey Agar without crystal violet is less selective than MacConkey Agar, allowing it to permit the growth of staphylococci and enterococci.
- Permits growth of staphylococci and enterococci
- Intended to be used in the making of prepared media products
Recommended for the detection of coliforms and enteric pathogens in water, waste water, and foods, as well as for differentiating Mycobacterium.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together