Neutralizing Buffer, Hardy Diagnostics
Neutralizing Buffer, 10 ml, 16×125 mm tube, Pack of 20
Supplier: Hardy Diagnostics
Fournisseur #: K105
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Neutralizing Buffer allows the bacteriostatic effects of chlorine and quaternary ammonia compounds
- Allow sensitive organisms to thrive
- Neutralizes the bacteriostatic effects of chlorine
Neutralizes the bacteriostatic effects of chlorine and quaternary ammonia compounds, allowing sensitive organisms to thrive.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:Neutralizing Buffer
- Size:Pack of 20
- Format:16×125 mm tube
- Volume:10 ml
Specifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



