ContactUsLinkComponent
Egg Yolk Agar, Modified, Hardy Diagnostics
Egg Yolk Agar, Modified, Hardy Diagnostics
  75813-978
Product available on GSA Advantage®
 :  
undefined
Egg Yolk Agar, Modified, Hardy Diagnostics
  75813-978
 :  G215
Product available on GSA Advantage®
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Egg Yolk Agar, Mod.
  • Size:
    Pack of 10
  • Environmentally Preferable:
  • Minimum Order Quantity:
    10
  • Cat. no.:
    75813-978
  • Format:
    15×100 mm plate
  • Supplier No.:
    G215
Egg Yolk Agar, Modified, for Clostridium is used for the isolation, cultivation, and differentiation of Clostridium spp.
  • Isolation
  • Cultivation
  • Differentiation
For the isolation, cultivation, and differentiation of Clostridium spp. and other anaerobic bacteria.
 : Minimum order of 10 packs.